5,7-Dichloroimidazo[1,2-c]pyrimidine 95%
Dodavatel: Apollo Scientific
OR946694-1G
OR946694-250MG
APOSOR946694-1GEA
7280
CZK
APOSOR946694-1G
APOSOR946694-250MG
5,7-Dichloroimidazo[1,2-c]pyrimidine 95%
5,7-Dichloroimidazo[1,2-c]pyrimidine
Vzorec:
C₆H₃Cl₂N₃ MW: 188,014 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189631 CAS číslo: 85989-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...