2-(Pyrimidin-4-yl)acetonitrile 95%
Dodavatel: Apollo Scientific
OR946604-1G
OR946604-250MG
APOSOR946604-1GEA
18420
CZK
APOSOR946604-1G
APOSOR946604-250MG
2-(Pyrimidin-4-yl)acetonitrile 95%
2-(Pyrimidin-4-yl)acetonitrile
Vzorec:
C₆H₅N₃ Storage Temperature: Ambient |
MDL Number:
MFCD17015870 CAS číslo: 794522-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...