7-Benzyl-1,7-diazaspiro[4.5]decane ≥98%
Dodavatel: Apollo Scientific
OR946490-100MG
OR946490-1G
OR946490-500MG
APOSOR946490-100MGEA
16670
CZK
APOSOR946490-100MG
APOSOR946490-1G
APOSOR946490-500MG
7-Benzyl-1,7-diazaspiro[4.5]decane ≥98%
7-Benzyl-1,7-diazaspiro[4.5]decane
Vzorec:
C₁₅H₂₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD11223519 CAS číslo: 1086395-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...