4-Cyano-N-propylbenzamide 96%
Dodavatel: Apollo Scientific
OR946463-1G
OR946463-5G
APOSOR946463-1GEA
6220
CZK
APOSOR946463-1G
APOSOR946463-5G
4-Cyano-N-propylbenzamide 96%
4-Cyano-N-propylbenzamide
Vzorec:
C₁₁H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD09933204 CAS číslo: 855264-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...