Tisk…

2-Chloro-N-methyl-6-nitroaniline 98%

Dodavatel: Apollo Scientific

OR946450-10G OR946450-1G OR946450-5G
APOSOR946450-10GEA 14290 CZK
APOSOR946450-10G APOSOR946450-1G APOSOR946450-5G
2-Chloro-N-methyl-6-nitroaniline 98%
2-Chloro-N-methyl-6-nitroaniline
Vzorec: C₇H₇ClN₂O₂
MW: 186,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06408851
CAS číslo: 75438-12-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR