3-(Chloromethyl)-N,N-dimethylbenzamide 98%
Dodavatel: Apollo Scientific
OR946373-10G
OR946373-1G
OR946373-5G
APOSOR946373-10GEA
21490
CZK
APOSOR946373-10G
APOSOR946373-1G
APOSOR946373-5G
3-(Chloromethyl)-N,N-dimethylbenzamide 98%
3-(Chloromethyl)-N,N-dimethylbenzamide
Vzorec:
C₁₀H₁₂ClNO MW: 197,661 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11208394 CAS číslo: 442910-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...