1-Azaspiro[4.5]decane-2,8-dione
Dodavatel: Apollo Scientific
Synonyms:
1-azaspiro[4.5]decane-2,8-dione
OR946154-1G
OR946154-5G
APOSOR946154-1GEA
7510
CZK
APOSOR946154-1G
APOSOR946154-5G
1-Azaspiro[4.5]decane-2,8-dione
1-Azaspiro[4.5]decane-2,8-dione
Vzorec:
C₉H₁₃NO₂ MW: 167,205 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14706827 CAS číslo: 749861-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...