Tisk…

5-Chloro-2-methylpyrimidine-4-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

OR945918-1G OR945918-250MG
APOSOR945918-1GEA 23970 CZK
APOSOR945918-1G APOSOR945918-250MG
5-Chloro-2-methylpyrimidine-4-carboxylic acid 95%
5-Chloro-2-methylpyrimidine-4-carboxylic acid
Vzorec: C₆H₅ClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00223741
CAS číslo: 74840-47-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR