Tisk…

N,N-Dimethyl-1H-pyrazolo[3,4-b]pyridin-6-amine 95%

Dodavatel: Apollo Scientific

OR945862-1G OR945862-5G
APOSOR945862-1GEA 4630 CZK
APOSOR945862-1G APOSOR945862-5G
N,N-Dimethyl-1H-pyrazolo[3,4-b]pyridin-6-amine 95%
N,N-Dimethyl-1H-pyrazolo[3,4-b]pyridin-6-amine
Vzorec: C₈H₁₀N₄
MW: 162,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13178167
CAS číslo: 63725-50-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR