Tisk…
Methyl 1-ethyl-1H-pyrazole-5-carboxylate 95%
Dodavatel: Apollo Scientific
Synonyms: Methyl 1-Ethyl-1H-pyrazole-5-carboxylate
OR945550-1G OR945550-250MG
APOSOR945550-1GEA 12730 CZK
APOSOR945550-1G APOSOR945550-250MG
Methyl 1-ethyl-1H-pyrazole-5-carboxylate 95%
Methyl 1-ethyl-1H-pyrazole-5-carboxylate
Vzorec: C₇H₁₀N₂O₂
MW: 154,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13174882
CAS číslo: 119458-46-7