Tisk…
Methyl 2-aminopent-4-enoate hydrochloride 95%
Dodavatel: Apollo Scientific
OR945414-1G OR945414-250MG
APOSOR945414-1GEA 3310 CZK
APOSOR945414-1G APOSOR945414-250MG
Methyl 2-aminopent-4-enoate hydrochloride 95%
Methyl 2-aminopent-4-enoate hydrochloride
Vzorec: C₆H₁₁NO₂·ClH
MW: 165,618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25955960
CAS číslo: 115289-55-9