3-Amino-5-chloropyrazine-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR945353-100MG
OR945353-250MG
APOSOR945353-100MGEA
4100
CZK
APOSOR945353-100MG
APOSOR945353-250MG
3-Amino-5-chloropyrazine-2-carboxylic acid 95%
3-Amino-5-chloropyrazine-2-carboxylic acid
Vzorec:
C₅H₄ClN₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD18250527 CAS číslo: 1260663-68-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...