Tisk…

3-Prop-2-ynoxybenzaldehyde 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Prop-2-yn-1-yloxy)benzaldehyde

OR945348-1G OR945348-250MG OR945348-5G
APOSOR945348-1GEA 827 CZK
APOSOR945348-1G APOSOR945348-250MG APOSOR945348-5G
3-Prop-2-ynoxybenzaldehyde 98%
3-Prop-2-ynoxybenzaldehyde
Vzorec: C₁₀H₈O₂
MW: 160,169 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05625818
CAS číslo: 5651-87-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR