Tisk…
2-(Methoxymethyl)-morpholine hydrochloride 97%
Dodavatel: Apollo Scientific
Synonyms: 2-(Methoxymethyl)morpholine hydrochloride
OR945273-1G OR945273-250MG OR945273-500MG
APOSOR945273-1GEA 27910 CZK
APOSOR945273-1G APOSOR945273-250MG APOSOR945273-500MG
2-(Methoxymethyl)-morpholine hydrochloride 97%
2-(Methoxymethyl)-morpholine hydrochloride
Vzorec: C₆H₁₃NO₂·ClH
MW: 167.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294690
CAS číslo: 144053-99-6