2-(1H-Indol-1-yl)propanoic acid 95%
Dodavatel: Apollo Scientific
OR945240-1G
OR945240-250MG
APOSOR945240-1GEA
25000
CZK
APOSOR945240-1G
APOSOR945240-250MG
2-(1H-Indol-1-yl)propanoic acid 95%
2-(1H-Indol-1-yl)propanoic acid
Vzorec:
C₁₁H₁₁NO₂ MW: 189,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11844460 CAS číslo: 131488-64-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...