Methyl 2-amino-2-phenylacetate
Dodavatel: Apollo Scientific
|
Varování
|
OR945143-1G
OR945143-5G
APOSOR945143-1GEA
3740
CZK
APOSOR945143-1G
APOSOR945143-5G
Methyl 2-amino-2-phenylacetate
Methyl 2-amino-2-phenylacetate
Vzorec:
C₉H₁₁NO₂ Storage Temperature: Freezer |
MDL Number:
MFCD01026128 CAS číslo: 26682-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...