Tisk…

6-Methoxy-3-methylisoquinoline 95%

Dodavatel: Apollo Scientific

OR945035-1G OR945035-250MG
APOSOR945035-1GEA 2730 CZK
APOSOR945035-1G APOSOR945035-250MG
6-Methoxy-3-methylisoquinoline 95%
6-Methoxy-3-methylisoquinoline
Vzorec: C₁₁H₁₁NO
MW: 173,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01846013
CAS číslo: 14446-31-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR