Tisk…

(4-Chloro-3-methoxyphenyl)methanamine 95%

Dodavatel: Apollo Scientific

OR944953-1G OR944953-250MG
APOSOR944953-1GEA 17990 CZK
APOSOR944953-1G APOSOR944953-250MG
(4-Chloro-3-methoxyphenyl)methanamine 95%
(4-Chloro-3-methoxyphenyl)methanamine
Vzorec: C₈H₁₀ClNO
MW: 171,624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09752213
CAS číslo: 247569-42-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR