Tisk…

2,3-Dimethyl-p-benzoquinone 95%

Dodavatel: Apollo Scientific

Synonyms: 2,3-Dimethylcyclohexa-2,5-diene-1,4-dione

OR944683-1G OR944683-250MG
APOSOR944683-1GEA 11810 CZK
APOSOR944683-1G APOSOR944683-250MG
2,3-Dimethyl-p-benzoquinone 95%
2,3-Dimethyl-p-benzoquinone
Vzorec: C₈H₈O₂
MW: 136,148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01663149
CAS číslo: 526-86-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR