1-Boc-2-acetyl-pyrrolidine 97%
Dodavatel: Apollo Scientific
OR944671-1G
OR944671-500MG
APOSOR944671-1GEA
13390
CZK
APOSOR944671-1G
APOSOR944671-500MG
1-Boc-2-acetyl-pyrrolidine 97%
1-Boc-2-acetyl-pyrrolidine
Vzorec:
C₁₁H₁₉NO₃ MW: 213,273 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16308624 CAS číslo: 92012-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...