3-Acetyl-3-chloro-dihydro-furan-2-one 95%
Dodavatel: Apollo Scientific
OR944586-1G
OR944586-250MG
APOSOR944586-1GEA
8270
CZK
APOSOR944586-1G
APOSOR944586-250MG
3-Acetyl-3-chloro-dihydro-furan-2-one 95%
3-Acetyl-3-chloro-dihydro-furan-2-one
Vzorec:
C₆H₇ClO₃ MW: 162,571 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08448079 CAS číslo: 2986-00-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...