Tisk…

Pyrazole-1-carbonyl chloride 95%

Dodavatel: Apollo Scientific

Synonyms: 1H-Pyrazole-1-carbonyl chloride

OR944313-1G OR944313-250MG
APOSOR944313-1GEA 22950 CZK
APOSOR944313-1G APOSOR944313-250MG
Pyrazole-1-carbonyl chloride 95%
Pyrazole-1-carbonyl chloride
Vzorec: C₄H₃ClN₂O
MW: 130,532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23378466
CAS číslo: 53355-55-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR