4,6-Dimethylpicolinic acid hydrochloride 95%
Dodavatel: Apollo Scientific
OR944312-1G
OR944312-250MG
APOSOR944312-1GEA
18420
CZK
APOSOR944312-1G
APOSOR944312-250MG
4,6-Dimethylpicolinic acid hydrochloride 95%
4,6-Dimethylpicolinic acid hydrochloride
Vzorec:
C₈H₉NO₂·ClH MW: 187,624 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23378465 CAS číslo: 1391733-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...