Tisk…
(1-Cyclopropyl-1-methylethyl)amine hydrochloride 95%
Dodavatel: Apollo Scientific
Synonyms: 2-Cyclopropylpropan-2-amine hydrochloride
OR944298-1G OR944298-250MG
APOSOR944298-1GEA 1540 CZK
APOSOR944298-1G APOSOR944298-250MG
(1-Cyclopropyl-1-methylethyl)amine hydrochloride 95%
(1-Cyclopropyl-1-methylethyl)amine hydrochloride
Vzorec: C₆H₁₃N·ClH
MW: 135,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23144031
CAS číslo: 17397-13-6