Tisk…

5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 3-acetyl-1H-pyrazole-5-carboxylate

OR944243-1G
APOSOR944243-1GEA 3580 CZK
APOSOR944243-1G
5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester 95%
5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester
Vzorec: C₈H₁₀N₂O₃
MW: 182,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21362351
CAS číslo: 37622-89-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR