Tisk…

6-Bromo-1-methoxyisoquinoline 95%

Dodavatel: Apollo Scientific
Varování

OR944104-1G OR944104-250MG
APOSOR944104-1GEA 32240 CZK
APOSOR944104-1G APOSOR944104-250MG
6-Bromo-1-methoxyisoquinoline 95%
6-Bromo-1-methoxyisoquinoline
Vzorec: C₁₀H₈BrNO
MW: 238,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16877645
CAS číslo: 891785-23-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR