Tisk…

3-(Methoxymethyl)-pyrrolidine hydrochloride 97%

Dodavatel: Apollo Scientific

Synonyms: 3-(Methoxymethyl)pyrrolidine hydrochloride

OR943851-1G
APOSOR943851-1GEA 2110 CZK
APOSOR943851-1G
3-(Methoxymethyl)-pyrrolidine hydrochloride 97%
3-(Methoxymethyl)-pyrrolidine hydrochloride
Vzorec: C₆H₁₃NO·ClH
MW: 151,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997928
CAS číslo: 1185303-79-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR