Tisk…

N,N-Dimethyl-2-morpholinemethanamine dihydrochloride 97%

Dodavatel: Apollo Scientific

OR943850-1G OR943850-250MG OR943850-500MG
APOSOR943850-1GEA 18620 CZK
APOSOR943850-1G APOSOR943850-250MG APOSOR943850-500MG
N,N-Dimethyl-2-morpholinemethanamine dihydrochloride 97%
N,N-Dimethyl-2-morpholinemethanamine dihydrochloride
Vzorec: C₇H₁₆N₂O·2ClH
MW: 217,137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997816
CAS číslo: 122894-40-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR