Tisk…

(8-Methylimidazo[1,2-a]pyridin-3-yl)methanol 95%

Dodavatel: Apollo Scientific

OR943822-1G OR943822-5G
APOSOR943822-1GEA 7250 CZK
APOSOR943822-1G APOSOR943822-5G
(8-Methylimidazo[1,2-a]pyridin-3-yl)methanol 95%
(8-Methylimidazo[1,2-a]pyridin-3-yl)methanol
Vzorec: C₉H₁₀N₂O
MW: 162,188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864458
CAS číslo: 178488-40-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR