Tisk…

1-(Thiophen-2-yl)ethan-1-ol 95%

Dodavatel: Apollo Scientific

OR943811-1G OR943811-250MG
APOSOR943811-1GEA 18420 CZK
APOSOR943811-1G APOSOR943811-250MG
1-(Thiophen-2-yl)ethan-1-ol 95%
1-(Thiophen-2-yl)ethan-1-ol
Vzorec: C₆H₈OS
MW: 128,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836745
CAS číslo: 2309-47-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR