Tisk…

N,5-Dimethyl-1,3,4-thiadiazol-2-amine 95%

Dodavatel: Apollo Scientific

OR943597-1G OR943597-250MG
APOSOR943597-1GEA 22950 CZK
APOSOR943597-1G APOSOR943597-250MG
N,5-Dimethyl-1,3,4-thiadiazol-2-amine 95%
N,5-Dimethyl-1,3,4-thiadiazol-2-amine
Vzorec: C₄H₇N₃S
MW: 129,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00986507
CAS číslo: 38917-35-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR