Tisk…

7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile 95%

Dodavatel: Apollo Scientific

Synonyms: 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile

OR943560-1G OR943560-250MG
APOSOR943560-1GEA 18420 CZK
APOSOR943560-1G APOSOR943560-250MG
7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile 95%
7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile
Vzorec: C₈H₇N₅
MW: 173,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00206563
CAS číslo: 255389-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR