Tisk…

(1S)-1-(3-Nitrophenyl)ethanamine 95%

Dodavatel: Apollo Scientific

OR943498-1G OR943498-250MG
APOSOR943498-1GEA 23580 CZK
APOSOR943498-1G APOSOR943498-250MG
(1S)-1-(3-Nitrophenyl)ethanamine 95%
(1S)-1-(3-Nitrophenyl)ethanamine
Vzorec: C₈H₁₀N₂O₂
MW: 166,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659477
CAS číslo: 297730-25-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR