N,N-Dimethyl-3-morpholinemethanamine dihydrochloride 97%
Dodavatel: Apollo Scientific
OR943483-100MG
OR943483-250MG
OR943483-500MG
APOSOR943483-100MGEA
8070
CZK
APOSOR943483-100MG
APOSOR943483-250MG
APOSOR943483-500MG
N,N-Dimethyl-3-morpholinemethanamine dihydrochloride 97%
N,N-Dimethyl-3-morpholinemethanamine dihydrochloride
Vzorec:
C₇H₁₆N₂O·2ClH MW: 217,137 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31560278 CAS číslo: 128454-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...