Tisk…

1-(6-Methoxy-2,3-dihydro-1H-inden-5-yl)methanamine 95%

Dodavatel: Apollo Scientific

OR943370-1G OR943370-250MG
APOSOR943370-1GEA 21490 CZK
APOSOR943370-1G APOSOR943370-250MG
1-(6-Methoxy-2,3-dihydro-1H-inden-5-yl)methanamine 95%
1-(6-Methoxy-2,3-dihydro-1H-inden-5-yl)methanamine
Vzorec: C₁₁H₁₅NO
MW: 177,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07403242
CAS číslo: 879063-08-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR