Tisk…

6-Methyl-benzo[d]isoxazol-3-ylamine 95%

Dodavatel: Apollo Scientific

Synonyms: 6-Methylbenzo[d]isoxazol-3-amine

OR943182-1G OR943182-250MG
APOSOR943182-1GEA 19150 CZK
APOSOR943182-1G APOSOR943182-250MG
6-Methyl-benzo[d]isoxazol-3-ylamine 95%
6-Methyl-benzo[d]isoxazol-3-ylamine
Vzorec: C₈H₈N₂O
MW: 148,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18803474
CAS číslo: 1378699-89-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR