Tisk…
2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid ethyl ester 95%
Dodavatel: Apollo Scientific
OR943129-1G OR943129-250MG
APOSOR943129-1GEA 20900 CZK
APOSOR943129-1G APOSOR943129-250MG
2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid ethyl ester 95%
2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid ethyl ester
Vzorec: C₉H₁₂N₂O₃
MW: 196,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26593859
CAS číslo: 1297547-66-0