6-Bromo-N-methylpyridin-3-amine 95%
Dodavatel: Apollo Scientific
OR943102-5G
APOSOR943102-5GEA
3380
CZK
APOSOR943102-5G
6-Bromo-N-methylpyridin-3-amine 95%
6-Bromo-N-methylpyridin-3-amine
Vzorec:
C₆H₇BrN₂ MW: 187,037 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234191 CAS číslo: 1022641-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...