2-Chloro-6-methylamino-benzoic acid methyl ester 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 2-chloro-6-(methylamino)benzoate
OR943100-1G
OR943100-250MG
APOSOR943100-1GEA
7910
CZK
APOSOR943100-1G
APOSOR943100-250MG
2-Chloro-6-methylamino-benzoic acid methyl ester 95%
2-Chloro-6-methylamino-benzoic acid methyl ester
Vzorec:
C₉H₁₀ClNO₂ MW: 199.634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24387373 CAS číslo: 1379595-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...