3-Oxaspiro[5.5]undecan-9-one
Dodavatel: Apollo Scientific
OR942976-1G
OR942976-250MG
APOSOR942976-1GEA
22250
CZK
APOSOR942976-1G
APOSOR942976-250MG
3-Oxaspiro[5.5]undecan-9-one
3-Oxaspiro[5.5]undecan-9-one
Vzorec:
C₁₀H₁₆O₂ MW: 168,233 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17013084 CAS číslo: 1159280-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...