Tisk…

3-Chloro-2-methylbenzaldehyde 95%

Dodavatel: Apollo Scientific

OR942967-1G OR942967-5G
APOSOR942967-1GEA 2530 CZK
APOSOR942967-1G APOSOR942967-5G
3-Chloro-2-methylbenzaldehyde 95%
3-Chloro-2-methylbenzaldehyde
Vzorec: C₈H₇ClO
MW: 154,594 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11047716
CAS číslo: 874-27-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR