Tisk…

Methyl 3-amino-5-chlorobenzothiophene-2-carboxylate

Dodavatel: Apollo Scientific

OR942899-1G OR942899-250MG
APOSOR942899-1GEA 8700 CZK
APOSOR942899-1G APOSOR942899-250MG
Methyl 3-amino-5-chlorobenzothiophene-2-carboxylate
Methyl 3-amino-5-chlorobenzothiophene-2-carboxylate
Vzorec: C₁₀H₈ClNO₂S
MW: 241,694 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13689054
CAS číslo: 1026241-99-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR