Methyl 2-chloro-6-hydroxybenzoate 95%
Dodavatel: Apollo Scientific
OR942749-1G
OR942749-250MG
APOSOR942749-1GEA
3020
CZK
APOSOR942749-1G
APOSOR942749-250MG
Methyl 2-chloro-6-hydroxybenzoate 95%
Methyl 2-chloro-6-hydroxybenzoate
Vzorec:
C₈H₇ClO₃ MW: 186,592 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15523524 CAS číslo: 176750-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...