Methyl 3-chloro-2,2-dimethylbut-3-enoate ≥98%
Dodavatel: Apollo Scientific
OR942701-100MG
OR942701-1G
OR942701-250MG
APOSOR942701-100MGEA
11050
CZK
APOSOR942701-100MG
APOSOR942701-1G
APOSOR942701-250MG
Methyl 3-chloro-2,2-dimethylbut-3-enoate ≥98%
Methyl 3-chloro-2,2-dimethylbut-3-enoate
Vzorec:
C₇H₁₁ClO₂ MW: 162,614 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093339 CAS číslo: 56663-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...