1-(4-Hydroxyphenyl)cyclopropane-1-carbonitrile 98%
Dodavatel: Apollo Scientific
OR942695-1G
OR942695-250MG
APOSOR942695-1GEA
18620
CZK
APOSOR942695-1G
APOSOR942695-250MG
1-(4-Hydroxyphenyl)cyclopropane-1-carbonitrile 98%
1-(4-Hydroxyphenyl)cyclopropane-1-carbonitrile
Vzorec:
C₁₀H₉NO MW: 159,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19692083 CAS číslo: 1073477-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...