2-Chloro-6-methylamino-benzonitrile 95%
Dodavatel: Apollo Scientific
OR942586-1G
OR942586-250MG
APOSOR942586-1GEA
13860
CZK
APOSOR942586-1G
APOSOR942586-250MG
2-Chloro-6-methylamino-benzonitrile 95%
2-Chloro-6-methylamino-benzonitrile
Vzorec:
C₈H₇ClN₂ MW: 166,608 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16663224 CAS číslo: 20925-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...