Methyl 2,4-dihydroxy-6-methylbenzoate 95%
Dodavatel: Apollo Scientific
OR942500-1G
OR942500-250MG
APOSOR942500-1GEA
11870
CZK
APOSOR942500-1G
APOSOR942500-250MG
Methyl 2,4-dihydroxy-6-methylbenzoate 95%
Methyl 2,4-dihydroxy-6-methylbenzoate
Vzorec:
C₉H₁₀O₄ MW: 182,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00184668 CAS číslo: 3187-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...