Tisk…

5-Chloro-2-(methylamino)phenol 95%

Dodavatel: Apollo Scientific

OR942472-1G OR942472-250MG
APOSOR942472-1GEA 16340 CZK
APOSOR942472-1G APOSOR942472-250MG
5-Chloro-2-(methylamino)phenol 95%
5-Chloro-2-(methylamino)phenol
Vzorec: C₇H₈ClNO
MW: 157.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16998610
CAS číslo: 1243357-05-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR