Tisk…

2-Bromo-3-hydroxybenzonitrile 95%

Dodavatel: Apollo Scientific

OR942456-1G OR942456-250MG
APOSOR942456-1GEA 6350 CZK
APOSOR942456-1G APOSOR942456-250MG
2-Bromo-3-hydroxybenzonitrile 95%
2-Bromo-3-hydroxybenzonitrile
Vzorec: C₇H₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD09266204
CAS číslo: 693232-06-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR