Tisk…

2-Amino-5-chloro-4-cyanopyridine 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-5-chloroisonicotinonitrile

OR942360-1G OR942360-250MG
APOSOR942360-1GEA 5200 CZK
APOSOR942360-1G APOSOR942360-250MG
2-Amino-5-chloro-4-cyanopyridine 95%
2-Amino-5-chloro-4-cyanopyridine
Vzorec: C₆H₄ClN₃
MW: 153.569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26792763
CAS číslo: 1393106-21-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR